Anti-EPOR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 38409]

Supplier: R & D Systems
FAB307P
220017-544EA 549.54 USD
220017-544
Anti-EPOR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 38409]
Antibodies
Type: Primary
Antigen: EPOR
Clonality: Monoclonal
Clone: 38409
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR