Anti-EPOR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 38421]

Supplier: R & D Systems
FAB3072A
102882-398EA 577.02 USD
102882-398
Anti-EPOR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 38421]
Antibodies
Type: Primary
Antigen: EPOR
Clonality: Monoclonal
Clone: 38421
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR