Anti-IL7R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 40131]

Supplier: R & D Systems
FAB306P-100 FAB306P-025
102882-394EA 268.98 USD
102882-394 102882-396
Anti-IL7R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 40131]
Antibodies
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: 40131
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR