Anti-ITGAV Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 23C6]

Supplier: R & D Systems
FAB3050A
220017-539EA 608.83 USD
220017-539
Anti-ITGAV Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 23C6]
Antibodies
Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone: 23C6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR