Anti-EPHB4 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 395810]

Supplier: R & D Systems
FAB3038N
102882-374EA 543.75 USD
102882-374
Anti-EPHB4 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 395810]
Antibodies
Type: Primary
Antigen: EPHB4
Clonality: Monoclonal
Clone: 395810
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR