Anti-EPHA2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 371805]

Supplier: R & D Systems
FAB3035G
102882-364EA 543.75 USD
102882-364
Anti-EPHA2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 371805]
Antibodies
Type: Primary
Antigen: EPHA2
Clonality: Monoclonal
Clone: 371805
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR