Anti-IL3RA Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 32703]

Supplier: R & D Systems
FAB301N
102882-358EA 501.82 USD
102882-358
Anti-IL3RA Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 32703]
Antibodies
Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone: 32703
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR