Anti-IL3RA Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 32703]

Supplier: R & D Systems
FAB301C
220017-535EA 543.75 USD
220017-535
Anti-IL3RA Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 32703]
Antibodies
Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone: 32703
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR