Anti-SDC2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 305515]

Supplier: R & D Systems
FAB2965A
220017-531EA 639.2 USD
220017-531
Anti-SDC2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 305515]
Antibodies
Type: Primary
Antigen: SDC2
Clonality: Monoclonal
Clone: 305515
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR