Anti-MARCO Rat Monoclonal Antibody (Fluorescein) [clone: 579511]

Supplier: R & D Systems
FAB2956F
220017-529EA 420.83 USD
220017-529
Anti-MARCO Rat Monoclonal Antibody (Fluorescein) [clone: 579511]
Antibodies
Type: Primary
Antigen: MARCO
Clonality: Monoclonal
Clone: 579511
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR