Anti-MARCO Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 579511]

Supplier: R & D Systems
FAB2956A
220017-528EA 600.15 USD
220017-528
Anti-MARCO Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 579511]
Antibodies
Type: Primary
Antigen: MARCO
Clonality: Monoclonal
Clone: 579511
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR