Anti-CLEC4G Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 845404]

Supplier: R & D Systems
FAB2947A
102882-340EA 600.15 USD
102882-340
Anti-CLEC4G Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 845404]
Antibodies
Type: Primary
Antigen: CLEC4G
Clonality: Monoclonal
Clone: 845404
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR