Anti-CLEC12A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 687317]

Supplier: R & D Systems
FAB2946P
102882-338EA 543.75 USD
102882-338
Anti-CLEC12A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 687317]
Antibodies
Type: Primary
Antigen: CLEC12A
Clonality: Monoclonal
Clone: 687317
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR