Anti-CLEC12A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 687317]

Supplier: R & D Systems
FAB2946A
102882-330EA 577.02 USD
102882-330
Anti-CLEC12A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 687317]
Antibodies
Type: Primary
Antigen: CLEC12A
Clonality: Monoclonal
Clone: 687317
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR