Anti-SDC4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 336304]

Supplier: R & D Systems
FAB29181A
102882-324EA 600.15 USD
102882-324
Anti-SDC4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 336304]
Antibodies
Type: Primary
Antigen: SDC4
Clonality: Monoclonal
Clone: 336304
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR