Anti-KIR3DL2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 539304]

Supplier: R & D Systems
FAB2878N-100 FAB2878N-025
102882-314EA 160.52 USD
102882-314 102882-316
Anti-KIR3DL2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 539304]
Antibodies
Type: Primary
Antigen: KIR3DL2
Clonality: Monoclonal
Clone: 539304
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR