Anti-SIGLEC6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 767329]

Supplier: R & D Systems
FAB2859P
102882-304EA 507.6 USD
102882-304
Anti-SIGLEC6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 767329]
Antibodies
Type: Primary
Antigen: SIGLEC6
Clonality: Monoclonal
Clone: 767329
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR