Anti-SIGLEC6 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 767329]

Supplier: R & D Systems
FAB2859G
102882-302EA 513.38 USD
102882-302
Anti-SIGLEC6 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 767329]
Antibodies
Type: Primary
Antigen: SIGLEC6
Clonality: Monoclonal
Clone: 767329
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR