Anti-SIGLEC6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 767329]

Supplier: R & D Systems
FAB2859A
102882-300EA 527.85 USD
102882-300
Anti-SIGLEC6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 767329]
Antibodies
Type: Primary
Antigen: SIGLEC6
Clonality: Monoclonal
Clone: 767329
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR