Anti-IL2RG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 633162]

Supplier: R & D Systems
FAB2842A
102882-246EA 577.02 USD
102882-246
Anti-IL2RG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 633162]
Antibodies
Type: Primary
Antigen: IL2RG
Clonality: Monoclonal
Clone: 633162
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR