Anti-CCR10 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 248918]

Supplier: R & D Systems
FAB2815N-025 FAB2815N-100
102882-240EA 566.89 USD
102882-240 102882-014
Anti-CCR10 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 248918]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 248918
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR