Anti-CCR10 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 248918]

Supplier: R & D Systems
FAB2815A-100 FAB2815A-025
102882-006EA 244.4 USD
102882-006 102882-008
Anti-CCR10 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 248918]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 248918
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR