Anti-CLEC4D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 413512]

Supplier: R & D Systems
FAB2806P
220017-520EA 501.82 USD
220017-520
Anti-CLEC4D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 413512]
Antibodies
Type: Primary
Antigen: CLEC4D
Clonality: Monoclonal
Clone: 413512
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR