Anti-ITGA3 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB2787A
220017-517EA 513.38 USD
220017-517
Anti-ITGA3 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: ITGA3
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR