Anti-CD300LF Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 234903]

Supplier: R & D Systems
FAB2774G
102882-230EA 513.38 USD
102882-230
Anti-CD300LF Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 234903]
Antibodies
Type: Primary
Antigen: CD300LF
Clonality: Monoclonal
Clone: 234903
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR