Anti-TDGF1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 89633]

Supplier: R & D Systems
FAB2772G
102882-226EA 561.11 USD
102882-226
Anti-TDGF1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 89633]
Antibodies
Type: Primary
Antigen: TDGF1
Clonality: Monoclonal
Clone: 89633
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR