Anti-IL31RA Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB2769P
220017-508EA 517.72 USD
220017-508
Anti-IL31RA Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: IL31RA
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR