Anti-HCAR2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 245106]

Supplier: R & D Systems
FAB2760A
220017-503EA 608.83 USD
220017-503
Anti-HCAR2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 245106]
Antibodies
Type: Primary
Antigen Symbol: HCAR2
Antigen Name: hydroxycarboxylic acid receptor 2
Antigen Synonyms: GPR109A, HCA2, HM74a, HM74b, NIACR1, PUMAG, Puma-g
Clonality: Monoclonal
Clone: 245106
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR