Anti-TLR4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 267518]

Supplier: R & D Systems
FAB2759P
220017-502EA 513.38 USD
220017-502
Anti-TLR4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 267518]
Antibodies
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 267518
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR