Anti-CD200 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 325516]

Supplier: R & D Systems
FAB27241P
220017-497EA 480.12 USD
220017-497
Anti-CD200 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 325516]
Antibodies
Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: 325516
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR