Anti-CD200 Mouse Monoclonal Antibody (Fluorescein) [clone: 325516]

Supplier: R & D Systems
FAB27241F
220017-496EA 308.03 USD
220017-496
Anti-CD200 Mouse Monoclonal Antibody (Fluorescein) [clone: 325516]
Antibodies
Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: 325516
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR