Anti-CDH6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 427909]

Supplier: R & D Systems
FAB2715P
220017-494EA 513.38 USD
220017-494
Anti-CDH6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 427909]
Antibodies
Type: Primary
Antigen: CDH6
Clonality: Monoclonal
Clone: 427909
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR