Anti-CDH6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 427909]

Supplier: R & D Systems
FAB2715A
220017-493EA 561.11 USD
220017-493
Anti-CDH6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 427909]
Antibodies
Type: Primary
Antigen: CDH6
Clonality: Monoclonal
Clone: 427909
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR