Anti-MSR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 351615]

Supplier: R & D Systems
FAB2708P
220017-492EA 543.75 USD
220017-492
Anti-MSR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 351615]
Antibodies
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 351615
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR