Anti-IL1R1 Goat Polyclonal Antibody (Alexa Fluor® 700)

Supplier: R & D Systems
FAB269N-025 FAB269N-100
102882-200EA 284.89 USD
102882-200 102882-202
Anti-IL1R1 Goat Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR