Anti-IL1R1 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB269F-025 FAB269F-100
102882-196EA 219.82 USD
102882-196 102882-198
Anti-IL1R1 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR