Anti-IL1R1 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB269A-025 FAB269A-100
102882-192EA 326.83 USD
102882-192 102882-194
Anti-IL1R1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR