Anti-LAIR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 342219]

Supplier: R & D Systems
FAB2664P
220017-486EA 561.11 USD
220017-486
Anti-LAIR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 342219]
Antibodies
Type: Primary
Antigen: LAIR1
Clonality: Monoclonal
Clone: 342219
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR