Anti-PVRL4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 337516]

Supplier: R & D Systems
FAB2659A
220017-482EA 582.8 USD
220017-482
Anti-PVRL4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 337516]
Antibodies
Type: Primary
Antigen: PVRL4
Clonality: Monoclonal
Clone: 337516
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR