Anti-CLEC4A2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 320507]

Supplier: R & D Systems
FAB26171P
220017-481EA 513.38 USD
220017-481
Anti-CLEC4A2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 320507]
Antibodies
Type: Primary
Antigen: CLEC4A2
Clonality: Monoclonal
Clone: 320507
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR