Anti-TLR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 383936]

Supplier: R & D Systems
FAB2616P
220017-480EA 527.85 USD
220017-480
Anti-TLR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 383936]
Antibodies
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: 383936
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR