Anti-ULBP1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 237104]

Supplier: R & D Systems
FAB2588A
102882-166EA 577.02 USD
102882-166
Anti-ULBP1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 237104]
Antibodies
Type: Primary
Antigen: ULBP1
Clonality: Monoclonal
Clone: 237104
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR