Anti-CSPG4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: LHM-2]

Supplier: R & D Systems
FAB2585C
102882-164EA 594.37 USD
102882-164
Anti-CSPG4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: LHM-2]
Antibodies
Type: Primary
Antigen: CSPG4
Clonality: Monoclonal
Clone: LHM-2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR