Anti-CD16 Mouse Monoclonal Antibody (Fluorescein) [clone: 245536]

Supplier: R & D Systems
FAB2546F-100 FAB2546F-025
102882-152EA 160.52 USD
102882-152 102882-154
Anti-CD16 Mouse Monoclonal Antibody (Fluorescein) [clone: 245536]
Antibodies
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 245536
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR