Anti-CD16 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 245536]

Supplier: R & D Systems
FAB2546C-100 FAB2546C-025
102882-148EA 160.52 USD
102882-148 102882-150
Anti-CD16 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 245536]
Antibodies
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 245536
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR