Anti-CD16 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 245536]

Supplier: R & D Systems
FAB2546A-025 FAB2546A-100
102882-144EA 179.32 USD
102882-144 102882-146
Anti-CD16 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 245536]
Antibodies
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 245536
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR