Anti-IL5RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 26815]

Supplier: R & D Systems
FAB253A
220017-460EA 549.54 USD
220017-460
Anti-IL5RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 26815]
Antibodies
Type: Primary
Antigen: IL5RA
Clonality: Monoclonal
Clone: 26815
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR