Anti-MRC1 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB2535P
102882-142EA 561.11 USD
102882-142
Anti-MRC1 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR