Anti-MRC1 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB2535G
102882-138EA 513.38 USD
102882-138
Anti-MRC1 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR