Anti-MRC1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 857615]

Supplier: R & D Systems
FAB25351A
102882-132EA 561.11 USD
102882-132
Anti-MRC1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 857615]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 857615
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR